Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099097.1 | NP_001092567.1 | 5 - 6 | 637 | 66 | |
GenBank mRNA | BC146167.1 | - | 5 - 6 | 637 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |