Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099117.1 | NP_001092587.1 | 1 - 2 | 113 | 77 | |
GenBank mRNA | BC142507.1 | - | 1 - 2 | 113 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |