Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOGAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099136.1 | NP_001092606.1 | 5 - 6 | 861 | 69 | |
GenBank mRNA | BC142148.1 | - | 5 - 6 | 861 | 69 | |
HM921049.1 | - | 5 - 6 | 850 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |