Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB6B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101129.1 | NP_001094599.1 | 5 - 6 | 528 | 106 | |
GenBank mRNA | BC150111.1 | - | 5 - 6 | 528 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |