Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL2L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101148.2 | NP_001094618.1 | 2 - 3 | 204 | 103 | |
GenBank mRNA | BC149358.1 | - | 2 - 3 | 205 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |