Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101184.1 | NP_001094654.1 | 4 - 5 | 856 | 69 | |
GenBank mRNA | BT030682.1 | - | 4 - 5 | 856 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |