Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101200.2 | NP_001094670.1 | 4 - 5 | 384 | 83 | |
GenBank mRNA | BC151255.1 | - | 4 - 5 | 385 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |