Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C26H10orf82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101253.2 | NP_001094723.1 | 5 - 6 | 597 | 68 | |
GenBank mRNA | BC149249.1 | - | 5 - 6 | 598 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |