Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102218.2 | NP_001095688.1 | 3 - 4 | 492 | 108 | |
GenBank mRNA | BC148102.1 | - | 3 - 4 | 495 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |