Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDYL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102223.2 | NP_001095693.1 | 6 - 7 | 1728 | 91 | |
GenBank mRNA | BC151603.1 | - | 6 - 7 | 1730 | 91 | |
EF687906.1 | - | 8 - 9 | 1811 | 91 | ||
EF690280.1 | - | 6 - 7 | 1781 | 91 | ||
EF690281.1 | - | 5 - 6 | 1114 | 91 | ||
EF690282.1 | - | 3 - 4 | 672 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |