Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZYG11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102247.1 | NP_001095717.1 | 7 - 8 | 1568 | 80 | |
GenBank mRNA | BC151363.1 | - | 7 - 8 | 1568 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |