Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102266.1 | NP_001095736.1 | 1 - 2 | 45 | 102 | |
GenBank mRNA | BC148038.1 | - | 1 - 2 | 45 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |