Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHROOM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102294.1 | NP_001095764.1 | 3 - 4 | 1215 | 68 | |
GenBank mRNA | BC151639.1 | - | 3 - 4 | 1215 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |