Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102309.2 | NP_001095779.1 | 7 - 8 | 973 | 69 | |
GenBank mRNA | BC149934.1 | - | 7 - 8 | 974 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |