Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MCUR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102309.2 | NP_001095779.1 | 8 - 9 | 1081 | 78 | |
GenBank mRNA | BC149934.1 | - | 8 - 9 | 1082 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |