Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102323.1 | NP_001095793.1 | 18 - 19 | 2206 | 54 | |
GenBank mRNA | BC148934.1 | - | 18 - 19 | 2206 | 54 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |