Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYRFL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102323.1 | NP_001095793.1 | 6 - 7 | 902 | 73 | |
GenBank mRNA | BC148934.1 | - | 6 - 7 | 902 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |