Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2D2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105401.2 | NP_001098871.1 | 2 - 3 | 259 | 113 | |
GenBank mRNA | BC153239.1 | - | 2 - 3 | 260 | 113 | |
BT026286.1 | - | 1 - 2 | 156 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |