Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FNDC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105421.1 | NP_001098891.1 | 1 - 2 | 518 | 81 | |
GenBank mRNA | BC149873.1 | - | 1 - 2 | 518 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |