Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OXLD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105635.1 | NP_001099105.1 | 1 - 2 | 73 | 106 | |
GenBank mRNA | BC123903.1 | - | 1 - 2 | 73 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |