Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EWSR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109800.2 | NP_001103270.1 | 7 - 8 | 853 | 95 | |
GenBank mRNA | BC149284.1 | - | 7 - 8 | 817 | 95 | |
BC153844.1 | - | 7 - 8 | 854 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |