Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110803.2 | NP_001104273.2 | 4 - 5 | 893 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |