Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111104.1 | NP_001104574.1 | 8 - 9 | 1395 | 68 | |
GenBank mRNA | BC134633.1 | - | 8 - 9 | 1395 | 68 | |
EE371127.1 | - | 4 - 5 | 521 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |