Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CMC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113175.2 | NP_001106646.1 | 1 | 314 | 66 | |
GenBank mRNA | BC111685.1 | - | 1 | 315 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |