Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NKG2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098163.1 | NP_001091632.1 | 1 - 2 | 66 | 107 | |
GenBank mRNA | EF081289.1 | - | 1 - 2 | 66 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Bos taurus Probe spans exons |