Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098166.2 | NP_001091635.1 | 14 - 15 | 1552 | 136 | |
GenBank mRNA | BC140615.1 | - | 14 - 15 | 1553 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |