Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGFL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099187.2 | NP_001092657.1 | 8 - 9 | 919 | 115 | |
GenBank mRNA | BC146141.1 | - | 8 - 9 | 920 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |