Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H2orf73 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101283.2 | NP_001094753.1 | 3 - 4 | 453 | 74 | |
GenBank mRNA | BC149887.1 | - | 3 - 4 | 454 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |