Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CXCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101285.1 | NP_001094755.1 | 1 - 2 | 88 | 121 | |
GenBank mRNA | DQ328664.1 | - | 1 - 2 | 89 | 121 | |
EF634483.1 | - | 1 - 2 | 88 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Bos taurus Inventoried Probe spans exons |