Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HACD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103316.1 | NP_001096786.1 | 9 - 10 | 995 | 121 | |
GenBank mRNA | BC151339.1 | - | 9 - 10 | 995 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |