Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCTD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103341.1 | NP_001096811.1 | 1 - 2 | 279 | 82 | |
GenBank mRNA | BC142481.1 | - | 1 - 2 | 279 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |