Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105024.1 | NP_001098494.1 | 3 - 4 | 442 | 98 | |
GenBank mRNA | BC149085.1 | - | 3 - 4 | 442 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |