Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERPIND1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105046.2 | NP_001098516.1 | 4 - 5 | 1388 | 61 | |
GenBank mRNA | BC149414.1 | - | 4 - 5 | 1389 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |