Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105492.1 | NP_001098962.1 | 3 - 4 | 220 | 81 | |
GenBank mRNA | BC141981.1 | - | 3 - 4 | 348 | 81 | |
BC151447.1 | - | 3 - 4 | 220 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |