Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C18H16orf70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105502.1 | NP_001098972.1 | 9 - 10 | 766 | 78 | |
GenBank mRNA | BC142370.1 | - | 9 - 10 | 766 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |