Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105642.1 | NP_001099112.1 | 6 - 7 | 812 | 114 | |
GenBank mRNA | BC147884.1 | - | 6 - 7 | 812 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |