Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IZUMO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001110097.1 | NP_001103567.1 | 1 - 2 | 199 | 73 | |
GenBank mRNA | BC149306.1 | - | 1 - 2 | 199 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |