Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113572.1 | NP_001107044.1 | 8 - 9 | 1037 | 103 | |
GenBank mRNA | CN438393.1 | - | 1 - 2 | 103 | ||
DN743221.1 | - | 1 - 2 | 255 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Probe spans exons |