Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3-3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014389.2 | NP_001014411.1 | 1 | 92 | 134 | |
GenBank mRNA | AB099028.1 | - | 1 - 2 | 60 | 134 | |
BC134734.1 | - | 1 | 92 | 134 | ||
BT020962.1 | - | 1 - 2 | 75 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Both primers and probe map within a single exon |