Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176615.2 | NP_788788.1 | 7 - 8 | 928 | 132 | |
GenBank mRNA | AF020506.1 | - | 7 - 8 | 928 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |