Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176627.1 | NP_788800.1 | 3 - 4 | 328 | 148 | |
GenBank mRNA | AB098938.1 | - | 3 - 4 | 366 | 148 | |
AF192334.1 | - | 3 - 4 | 328 | 148 | ||
BC134714.1 | - | 3 - 4 | 377 | 148 | ||
BC151448.1 | - | 3 - 4 | 377 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |