Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174411.2 | NP_776836.1 | 8 - 9 | 1020 | 160 | |
GenBank mRNA | AB098781.2 | - | 1 - 2 | 297 | 160 | |
BC134743.1 | - | 8 - 9 | 1049 | 160 | ||
M73962.1 | - | 8 - 9 | 1020 | 160 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |