Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177490.1 | NP_803456.1 | 1 - 2 | 92 | 155 | |
GenBank mRNA | AF032866.1 | - | 1 - 2 | 67 | 155 | |
D84097.1 | - | 1 - 2 | 92 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |