Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076190.1 | NP_001069658.1 | 1 | 953 | 138 | |
GenBank mRNA | BC110199.1 | - | 1 | 953 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |