Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078022.1 | NP_001071490.1 | 2 | 1946 | 100 | |
GenBank mRNA | BC120044.1 | - | 2 | 1946 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |