Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LHFPL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104984.2 | NP_001098454.1 | 3 | 850 | 83 | |
GenBank mRNA | BC149448.1 | - | 3 | 851 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |