Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANBAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114861.1 | NP_001108333.1 | 3 - 4 | 740 | 77 | |
GenBank mRNA | BC120343.1 | - | 3 - 4 | 740 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |