Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MATN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143866.1 | NP_001137338.1 | 1 - 2 | 141 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Bos taurus Probe spans exons |