Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C17H5orf52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145133.1 | NP_001138605.1 | 1 - 2 | 354 | 83 | |
GenBank mRNA | DT836058.1 | - | 1 - 2 | 293 | 83 | |
DT850195.1 | - | 1 - 2 | 311 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Bos taurus Probe spans exons |