Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C27H8orf48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145540.1 | NP_001139012.1 | 1 | 1249 | 85 | |
GenBank mRNA | EE222395.1 | - | 1 | 441 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Bos taurus Both primers and probe map within a single exon |